Cobalt octoate ∼65% in mineral spirits (12% Co)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
27-0449-250G
27-0449-1000G
27-0449-50G
100204-020EA
104.74
USD
100204-020
100209-876
100209-878
Cobalt octoate ∼65% in mineral spirits (12% Co)
Cobalt octoate
Formula:
C₁₆H₃₀CoO₄ Density: 1.01 Flash Pt: 104 °F |
MDL Number:
MFCD00072632 CAS Number: 136-52-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...