Iron(III) meso-tetraphenylporphine-µ-oxo dimer

Supplier: STREM CHEMICALS INC MS

Synonyms: Iron(III) meso-tetraphenylporphine-mu-oxo dimer, Meso-5,10,15,20-Tetraphenyl-21H,23H-porphineironu-oxodimer

26-3900-2G 26-3900-500MG
100209-252EA 1579.57 USD
100209-252 100200-442
Iron(III) meso-tetraphenylporphine-µ-oxo dimer
Iron(III) meso-tetraphenylporphine-µ-oxo dimer

Porphine

Formula: C₈₈H₅₆Fe₂N₈O
MW: 1353.13 g/mol
MDL Number: MFCD00058898
CAS Number: 12582-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR