1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene CTC-Q-PHOS ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: CTC-Q-PHOS, 1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene, QPhos, 1,2,3,4,5-Pentaphenyl-1'-(di-t-butylphosphino)ferrocene

26-3575-100MG 26-3575-500MG
100205-052EA 108.98 USD
100205-052 100205-054
1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene CTC-Q-PHOS ≥95%
1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene CTC-Q-PHOS

Metallocenes, Derivatives & Cp Precursors, Phosphine

Formula: C₄₈H₄₇FeP
MW: 710.706 g/mol
MDL Number: MFCD03788940
CAS Number: 312959-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR