Pentamethylcyclopentadienyliron dicarbonyl dimer ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
26-3525-5G
26-3525-1G
100209-864EA
460
USD
100209-864
100209-862
Pentamethylcyclopentadienyliron dicarbonyl dimer ≥98%
Pentamethylcyclopentadienyliron dicarbonyl dimer
Metallocenes, Derivatives & Cp Precursors
Formula:
C₂₄H₃₀Fe₂O₄ |
MDL Number:
MFCD00058862 CAS Number: 35344-11-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...