Iron(II) phthalocyanine ≥95%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Phthalocyanine Iron(II), Iron phthalocyanine
26-2825-1G
26-2825-5G
100205-048EA
42.46
USD
100205-048
100205-050
Iron(II) phthalocyanine ≥95%
Iron(II) phthalocyanine
Formula:
C₃₂H₁₆FeN₈ MW: 568.38 g/mol |
MDL Number:
MFCD00015953 CAS Number: 132-16-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...