Nonacarbonyldiiron ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Diiron nonacarbonyl, Iron nonacarbonyl

26-2640-25G 26-2640-5G 26-2640-100G
100206-516EA 256.18 USD
100206-516 100206-518 100206-514
Nonacarbonyldiiron ≥99%
Nonacarbonyldiiron

Metal Carbonyls

Formula: C₉Fe₂O₉
MW: 363.78 g/mol
Melting Pt: 100 °C (dec.)
Density: 2,85
MDL Number: MFCD00151465
CAS Number: 15321-51-4
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR