Iron(II) stearate (9% Fe)

Supplier: STREM CHEMICALS INC MS

26-2622-100G 26-2622-500G
100205-046EA 162.77 USD
100205-046 100204-018
Iron(II) stearate (9% Fe)
Iron(II) stearate
MDL Number: MFCD00050715
CAS Number: 2980-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR