Dodecacarbonyltriiron stabilized
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Triiron dodecacarbonyl, Iron dodecacarbonyl
Stabilization:
Stabilized with methanol 5-10%
26-2500-50G
26-2500-10G
100211-052EA
642.58
USD
100211-052
100206-510
Dodecacarbonyltriiron stabilized
Dodecacarbonyltriiron
Metal Carbonyls
Formula:
C₁₂Fe₃O₁₂ MW: 503.66 g/mol Melting Pt: 140 °C (dec.) Density: 2.00 |
MDL Number:
MFCD00135617 CAS Number: 17685-52-8 UN: 2926 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...