Iron sulfide ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Iron pyrite, Iron disulphide

26-2400-5000G 26-2400-1000G
100198-158EA 352.43 USD
100198-158 100198-728
Iron sulfide ≥95%
Iron sulfide
Formula: FeS₂
MW: 119.98 g/mol
MDL Number: MFCD00064690
CAS Number: 12068-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR