1-Hydroxyethylferrocene ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1-Ferrocenylethanol, (1-Hydroxyethyl)ferrocene, α-Hydroxyethylferrocene

26-2000-1G 26-2000-5G
100207-632EA 43.88 USD
100207-632 100205-044
1-Hydroxyethylferrocene ≥98%
1-Hydroxyethylferrocene
Formula: C₁₂H₅FeO
MW: 221.01 g/mol
Melting Pt: 76…77 °C
MDL Number: MFCD00013752
CAS Number: 1277-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR