1,1'-Ferrocenedicarboxylic acid ≥96%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
26-1800-5G
26-1800-1G
100202-384EA
358.09
USD
100202-384
100211-050
1,1'-Ferrocenedicarboxylic acid ≥96%
1,1'-Ferrocenedicarboxylic acid
Formula:
C₁₂H₁₀FeO₄ Melting Pt: 235 °C (dec.) |
MDL Number:
MFCD00001423 CAS Number: 1293-87-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...