Ethylferrocene ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

26-1600-25G 26-1600-5G
100196-928EA 493.97 USD
100196-928 100202-652
Ethylferrocene ≥98%
Ethylferrocene

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₂H₅Fe
Density: 1.256
MDL Number: MFCD00059071
CAS Number: 1273-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR