1,1'-Dimethylferrocene ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

26-1500-10G 26-1500-2G
100201-550EA 239.2 USD
100201-550 100201-552
1,1'-Dimethylferrocene ≥98%
1,1'-Dimethylferrocene

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₂H₁₄Fe
Melting Pt: 32…37 °C
MDL Number: MFCD00001425
CAS Number: 1291-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR