(Ferrocenylmethyl)trimethylammonium iodide ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: (Ferrocenylmethyl)trimethylammonium Iodide, Ferrocenylmethyltrimethylammonium iodide

26-1450-25G 26-1450-5G
100199-834EA 680.8 USD
100199-834 100211-046
(Ferrocenylmethyl)trimethylammonium iodide ≥99%
(Ferrocenylmethyl)trimethylammonium iodide
Formula: C₁₄H₂₀FeIN
MW: 385.07 g/mol
Melting Pt: 220 °C (dec.)
MDL Number: MFCD00041534
CAS Number: 12086-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR