(+/-)-N,N-Dimethyl-1-ferrocenylethylamine ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
26-1399-25G
26-1399-5G
26-1399-1G
100196-178EA
1803.2
USD
100196-178
100196-180
100200-668
(+/-)-N,N-Dimethyl-1-ferrocenylethylamine ≥98%
(+/-)-N,N-Dimethyl-1-ferrocenylethylamine
Metallocenes, Derivatives & Cp Precursors
Formula:
C₁₄H₁₉FeN Boiling Pt: 110 °C (0.45 mmHg) Density: 1,222 |
MDL Number:
MFCD01074444 CAS Number: 31904-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...