(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (2R)-1-[(1R)-1-[Bis(3,5-dimethylphenyl)phosphino]ethyl]-2-(diphenylphosphino)ferrocene

26-1255-100MG 26-1255-500MG
100209-844EA 157.11 USD
100209-844 100204-592
(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine ≥97%
(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi-3,5-xylylphosphine

JOSIPHOS, Phosphine

Formula: C₄₀H₄₀FeP₂
MDL Number: MFCD03426997
CAS Number: 184095-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR