(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldiphenylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenylethyl]diphenylphosphine

26-1230-100MG 26-1230-500MG
100209-842EA 157.11 USD
100209-842 100204-590
(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldiphenylphosphine ≥97%
(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldiphenylphosphine

JOSIPHOS, Phosphine

Formula: C₃₆H₄₄FeP₂
MW: 594.6000 g/mol
MDL Number: MFCD00800280
CAS Number: 158923-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR