(R)-(S)-Josiphos ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (2R)-1-[(1R)-1-(Dicyclohexylphosphino)ethyl]-2-(diphenylphosphino)ferrocene, (2Sp)-1-[(1R)-1-(Dicyclohexylphosphino)ethyl]-2-(diphenylphosphino)ferrocene, (R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine ethanol adduct, (R)-(-)-1-[(S)-2-Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine

26-1210-500MG 26-1210-100MG
100204-586EA 547.75 USD
100204-586 100209-838
(R)-(S)-Josiphos ≥97%
(R)-(S)-Josiphos

JOSIPHOS, Phosphine

Formula: C₃₆H₄₄FeP₂
MW: 594.541 g/mol
MDL Number: MFCD00800284
CAS Number: 155806-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR