(S)-(+)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethyldi-t-butylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (2S)-1-[(1S)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(diphenylphosphino)ferrocene

26-1201-500MG 26-1201-100MG
100204-584EA 547.75 USD
100204-584 100209-836
(S)-(+)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethyldi-t-butylphosphine ≥97%
(S)-(+)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethyldi-t-butylphosphine

JOSIPHOS, Phosphine

Formula: C₃₂H₄₀FeP₂
MDL Number: MFCD01074443
CAS Number: 277306-29-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR