(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi-t-butylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (2R)-1-[(1R)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(diphenylphosphino)ferrocene

26-1200-100MG 26-1200-500MG
100209-834EA 157.11 USD
100209-834 100204-582
(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi-t-butylphosphine ≥97%
(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldi-t-butylphosphine

JOSIPHOS, Phosphine

Formula: C₃₂H₄₀FeP₂
MW: 542.4500 g/mol
MDL Number: MFCD01074442
CAS Number: 155830-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR