(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]benzylamine ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

26-1151-100MG 26-1151-500MG
101418-774EA 229.29 USD
101418-774 101418-776
(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]benzylamine ≥98%
(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]benzylamine

Phosphine

MDL Number: MFCD08459348
CAS Number: 498580-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR