(R)-(-)-1-{(S)-2-[Bis(3,5-dimethyl-4-methoxyphenyl)phosphino]ferrocenyl}ethyldicyclohexylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (1R)-1-[Bis(4-methoxy-3,5-dimethylphenyl)phosphino]-2-[(1R)-1-(dicyclohexylphosphino)ethyl]ferrocene

26-1150-100MG 26-1150-500MG
100209-828EA 157.11 USD
100209-828 100204-576
(R)-(-)-1-{(S)-2-[Bis(3,5-dimethyl-4-methoxyphenyl)phosphino]ferrocenyl}ethyldicyclohexylphosphine ≥97%
(R)-(-)-1-{(S)-2-[Bis(3,5-dimethyl-4-methoxyphenyl)phosphino]ferrocenyl}ethyldicyclohexylphosphine

JOSIPHOS, Phosphine

Formula: C₄₂H₅₆FeO₂P₂
MW: 710.6900 g/mol
MDL Number: MFCD03426995
CAS Number: 360048-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR