(S)-(+)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldiphenylphosphine ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(S)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenylethyl]diphenylphosphine
26-1101-100MG
26-1101-500MG
100209-822EA
157.11
USD
100209-822
100204-570
(S)-(+)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldiphenylphosphine ≥97%
(S)-(+)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldiphenylphosphine
JOSIPHOS, Phosphine
Formula:
C₃₆H₄₄FeP₂ MW: 594.5900 g/mol |
MDL Number:
MFCD00800281 CAS Number: 162291-01-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...