(S)-(+)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldicyclohexylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS

Synonyms: (S)-1-[(S)-2-(Dicyclohexylphosphino)-ferrocenyl]ethyldicyclohexylphosphine

26-1001-500MG 26-1001-100MG
100204-568EA 547.75 USD
100204-568 100209-820
(S)-(+)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldicyclohexylphosphine ≥97%
(S)-(+)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldicyclohexylphosphine

JOSIPHOS, Phosphine

Formula: C₃₆H₅₆FeP₂
MW: 606.6400 g/mol
MDL Number: MFCD00800279
CAS Number: 246231-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR