(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldi-t-butylphosphine ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (2R)-1-[(1R)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(dicyclohexylphosphino)ferrocene, [(R)-1-[(R)-2-(Dicyclohexylphosphino)ferrocenyl]ethyl]di-tert-butylphosphine

26-0975-500MG 26-0975-100MG
100204-564EA 547.75 USD
100204-564 100209-816
(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldi-t-butylphosphine ≥97%
(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldi-t-butylphosphine

JOSIPHOS, Phosphine

Formula: C₃₂H₅₂FeP₂
MDL Number: MFCD08543435
CAS Number: 158923-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR