(R)-(+)-[(R)-2-Dicyclohexylphosphinoferrocenyl](N,N-dimethylamino)(2-dicyclohexylphosphinophenyl)methane ≥97%
Supplier: STREM CHEMICALS INC MS
Synonyms:
(1Rp)-1-(Dicyclohexylphosphino)-2-[(R)-[2-(dicyclohexylphosphino)phenyl](dimethylamino)methyl]ferrocene
26-0956-100MG
26-0956-500MG
101442-176EA
140.12
USD
101442-176
101442-178
(R)-(+)-[(R)-2-Dicyclohexylphosphinoferrocenyl](N,N-dimethylamino)(2-dicyclohexylphosphinophenyl)methane ≥97%
(R)-(+)-[(R)-2-Dicyclohexylphosphinoferrocenyl](N,N-dimethylamino)(2-dicyclohexylphosphinophenyl)methane
Phosphine
Formula:
C₄₃H₆₃FeNP₂ MW: 711.76 g/mol |
MDL Number:
MFCD04117706 CAS Number: 1156547-61-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...