(S)-(-)-[(S)-2-Dicyclohexylphosphinoferrocenyl](N,N-dimethylamino)(2-dicyclohexylphosphinophenyl)methane ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

26-0955-100MG 26-0955-500MG
100209-810EA 140.12 USD
100209-810 100204-558
(S)-(-)-[(S)-2-Dicyclohexylphosphinoferrocenyl](N,N-dimethylamino)(2-dicyclohexylphosphinophenyl)methane ≥97%
(S)-(-)-[(S)-2-Dicyclohexylphosphinoferrocenyl](N,N-dimethylamino)(2-dicyclohexylphosphinophenyl)methane

Phosphine

Formula: C₄₃H₆₃FeNP₂
MDL Number: MFCD04117706
CAS Number: 914089-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR