Cyclopentadienyliron dicarbonyl ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

26-0900-10G 26-0900-250G 26-0900-50G
100205-042EA 86.34 USD
100205-042 100211-042 100211-044
Cyclopentadienyliron dicarbonyl ≥99%
Cyclopentadienyliron dicarbonyl

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₄H₁₂Fe₂O₄
Melting Pt: 194 °C (dec.)
MDL Number: MFCD00001436
CAS Number: 12154-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR