Bis(i-propylcyclopentadienyl)iron ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

26-0450-5G 26-0450-1G
100202-382EA 888.86 USD
100202-382 100205-040
Bis(i-propylcyclopentadienyl)iron ≥98%
Bis(i-propylcyclopentadienyl)iron

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

MDL Number: MFCD01862456
CAS Number: 12126-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR