1,1'-Bis(diisopropylphosphino)ferrocene ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
DiPPF, 1,1'-Bis(di-i-propylphosphino)ferrocene
26-0275-500MG
26-0275-2G
100201-548EA
87.75
USD
100201-548
100206-506
1,1'-Bis(diisopropylphosphino)ferrocene ≥98%
1,1'-Bis(diisopropylphosphino)ferrocene
Phosphine
Formula:
C₂₂H₂₈FeP₂ MW: 410.25 g/mol |
MDL Number:
MFCD02684559 CAS Number: 97239-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...