1,1'-Bis(diphenylphosphino)ferrocene ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: DPPF, 1,1-Bis(diphenylphosphino)ferrocene

26-0270-5G 26-0270-1G
100196-174EA 145.78 USD
100196-174 100206-504
1,1'-Bis(diphenylphosphino)ferrocene ≥99%
1,1'-Bis(diphenylphosphino)ferrocene

Phosphine

Formula: C₃₄H₂₈FeP₂
MW: 554.39 g/mol
Melting Pt: 180 °C
MDL Number: MFCD00001422
CAS Number: 12150-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR