(S,S)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis(diphenylphosphino)ferrocene ≥97%
Supplier: STREM CHEMICALS INC MS
Synonyms:
(S,S)-(-)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis(diphenylphosphino)ferrocene
26-0252-500MG
26-0252-100MG
100204-554EA
518.03
USD
100204-554
100209-806
(S,S)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis(diphenylphosphino)ferrocene ≥97%
(S,S)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis(diphenylphosphino)ferrocene
MANDYPHOS, Phosphine
Formula:
C₅₂H₅₀FeN₂P₂ MW: 820.76 g/mol |
MDL Number:
MFCD04117705 CAS Number: 210842-74-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...