(S,S)-(-)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis[di(3,5-trifluoromethylphenyl)phosphino]ferrocene ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

26-0244-500MG 26-0244-100MG
100204-550EA 518.03 USD
100204-550 100209-802
(S,S)-(-)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis[di(3,5-trifluoromethylphenyl)phosphino]ferrocene ≥97%
(S,S)-(-)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis[di(3,5-trifluoromethylphenyl)phosphino]ferrocene

MANDYPHOS, Phosphine

MDL Number: MFCD04117703
CAS Number: 494227-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR