1,1'-Bis(di-tert-butylphosphino)ferrocene ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1,1'-Bis(di-t-butylphosphino)ferrocene, 1,1'-Bis[bis(1,1-dimethylethyl)phosphino]ferrocene, DTBPF

26-0150-2G 26-0150-500MG
100206-502EA 219.38 USD
100206-502 100201-546
1,1'-Bis(di-tert-butylphosphino)ferrocene ≥98%
1,1'-Bis(di-tert-butylphosphino)ferrocene

Phosphine

Formula: C₂₆H₄₄FeP₂
MW: 474.42 g/mol
MDL Number: MFCD01630818
CAS Number: 84680-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR