Manganese(III) acetate meso-tetraphenylporphine

Supplier: STREM CHEMICALS INC MS

Synonyms: Manganese(III) meso-tetraphenylporphine acetate

25-1470-1G 25-1470-5G
100205-032EA 246.28 USD
100205-032 100198-716
Manganese(III) acetate meso-tetraphenylporphine
Manganese(III) acetate meso-tetraphenylporphine

Porphine

Formula: C₄₆H₃₁MnN₄O₂
MW: 727.71 g/mol
MDL Number: MFCD00216548
CAS Number: 58356-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR