Manganese(II) phthalocyanine ≥97%

Supplier: STREM CHEMICALS INC MS

25-1460-5G 25-1460-1G
100206-500EA 409.05 USD
100206-500 100205-030
Manganese(II) phthalocyanine ≥97%
Manganese(II) phthalocyanine
Formula: C₃₂H₁₆MnN₈
MW: 567.46 g/mol
MDL Number: MFCD00049821
CAS Number: 14325-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR