Cyclopentadienylmanganese tricarbonyl ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cymantrene, Cyclopentadienylmanganese(I) tricarbonyl

25-0390-5G 25-0390-25G
100206-494EA 471.32 USD
100206-494 100206-492
Cyclopentadienylmanganese tricarbonyl ≥98%
Cyclopentadienylmanganese tricarbonyl

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₈H₅MnO₃
MW: 204.06 g/mol
Melting Pt: 77 °C
MDL Number: MFCD00001440
CAS Number: 12079-65-1
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR