(1S,2S)-(+)-[1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)]manganese(III) chloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S,S)-[N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediamine]manganese(III) chloride, (S,S)-Jacobsen Cat.

25-0301-5G 25-0301-1G
100203-056EA 267.51 USD
100203-056 100196-166
(1S,2S)-(+)-[1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)]manganese(III) chloride ≥98%
(1S,2S)-(+)-[1,2-Cyclohexanediamino-N,N'-bis(3,5-di-t-butylsalicylidene)]manganese(III) chloride

Jacobsen Ligand

Formula: C₃₆H₅₄Cl₃MnN₂O₂
MW: 708.1232 g/mol
Melting Pt: 325…326 °C
MDL Number: MFCD02101663
CAS Number: 135620-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR