(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
25-0300-5G
25-0300-1G
100203-054EA
257.6
USD
100203-054
100196-164
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride ≥98%
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride
Jacobsen Ligand
Formula:
C₃₆H₅₂ClMnN₂O₂ Melting Pt: 324…326 °C |
MDL Number:
MFCD02101664 CAS Number: 138124-32-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...