Bis(tetramethylcyclopentadienyl)manganese(II) ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Bis(tetramethylcyclopentadienyl)manganese

25-0265-1G 25-0265-5G
100203-052EA 206.65 USD
100203-052 100197-456
Bis(tetramethylcyclopentadienyl)manganese(II) ≥98%
Bis(tetramethylcyclopentadienyl)manganese (II)

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₈H₂₆Mn
MDL Number: MFCD01862454
CAS Number: 101932-75-6
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR