Bis(pentamethylcyclopentadienyl)manganese(II) ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Decamethylmanganocene, Bis(pentamethylcyclopentadienyl)manganese

25-0235-1G 25-0235-5G
100197-452EA 120.31 USD
100197-452 100197-454
Bis(pentamethylcyclopentadienyl)manganese(II) ≥98%
Bis(pentamethylcyclopentadienyl)manganese (II)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₂₀H₃₀Mn
MW: 325.39 g/mol
Melting Pt: 292 °C
MDL Number: MFCD00058844
CAS Number: 67506-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR