Bis(ethylcyclopentadienyl)manganese(II) ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Bis(ethylcyclopentadienyl)manganese

25-0210-5G 25-0210-1G
100202-376EA 612.86 USD
100202-376 100205-024
Bis(ethylcyclopentadienyl)manganese(II) ≥98%
Bis(ethylcyclopentadienyl)manganese (II)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₄H₁₈Mn
MW: 241.236 g/mol
MDL Number: MFCD01862452
CAS Number: 101923-26-6
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR