Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')chromium(III) ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)chromium(III), Cr(TMHD)3

24-1500-1G 24-1500-5G
100204-542EA 131.63 USD
100204-542 100206-100
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')chromium(III) ≥99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')chromium(III)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₃₃H₅₇CrO₆
MW: 601.8 g/mol
Boiling Pt: 270 °C (dec.)
Melting Pt: 230…232 °C
MDL Number: MFCD00058919
CAS Number: 14434-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR