Chromium(III) tetraphenylporphine chloride

Supplier: STREM CHEMICALS INC MS

24-0800-250MG 24-0800-1G
100206-490EA 284.49 USD
100206-490 100210-730
Chromium(III) tetraphenylporphine chloride
Chromium(III) tetraphenylporphine chloride

Jacobsen Ligand, Porphine

MDL Number: MFCD00269817
CAS Number: 28110-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR