Bis(tetramethylcyclopentadienyl)chromium(II) ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Bis(tetramethylcyclopentadienyl)chromium
24-0154-1G
24-0154-5G
100201-198EA
321.29
USD
100201-198
100197-016
Bis(tetramethylcyclopentadienyl)chromium(II) ≥98%
Bis(tetramethylcyclopentadienyl)chromium (II)
Metallocenes, Derivatives & Cp Precursors
Formula:
C₁₈H₂₆Cr MW: 294.40 g/mol |
MDL Number:
MFCD01862451 CAS Number: 82066-37-3 UN: 1325 ADR: 4.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...