Bis(pentamethylcyclopentadienyl)chromium(II) ≥95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Bis(pentamethylcyclopentadienyl)chromium, Decamethylchromocene
24-0150-1G
24-0150-5G
100210-800EA
309.97
USD
100210-800
100204-540
Bis(pentamethylcyclopentadienyl)chromium(II) ≥95%
Bis(pentamethylcyclopentadienyl)chromium (II)
Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD
Formula:
C₂₀H₃₀Cr MW: 322.45 g/mol |
MDL Number:
MFCD00058886 CAS Number: 74507-61-2 UN: 1325 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...