Bis(ethylcyclopentadienyl)chromium(II) ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Bis(ethylcyclopentadienyl)chromium
24-0147-1G
24-0147-5G
100207-370EA
225.05
USD
100207-370
100208-002
Bis(ethylcyclopentadienyl)chromium(II) ≥98%
Bis(ethylcyclopentadienyl)chromium (II)
Metallocenes, Derivatives & Cp Precursors
Formula:
C₁₄H₁₈Cr MW: 238.289 g/mol Boiling Pt: 95…98 °C (1.0 mmHg) Density: 1,15 |
MDL Number:
MFCD01862449 CAS Number: 55940-03-9 UN: 3186 ADR: 4.2,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...