Bis(cyclopentadienyl)chromium ≥95%, sublimated

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Chromocene, Bis(Cyclopentadienyl)Chromium(Ii)

24-0135-1G 24-0135-5G
100197-100EA 165.6 USD
100197-100 100207-244
Bis(cyclopentadienyl)chromium ≥95%, sublimated
Bis(cyclopentadienyl)chromium

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₀H₁₀Cr
MW: 182.19 g/mol
Melting Pt: 172…173 °C
MDL Number: MFCD00013750
CAS Number: 1271-24-5
UN: 3178
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR