Dibenzenechromium ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Bis(benzene)chromium

24-0130-5G 24-0130-1G
100201-540EA 653.91 USD
100201-540 100201-538
Dibenzenechromium ≥97%
Dibenzenechromium

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₂H₁₂Cr
MW: 208.22 g/mol
Melting Pt: 284…285 °C
MDL Number: MFCD00015485
CAS Number: 1271-54-1
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR