Chromium benzene tricarbonyl ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Benchrotrene, Benzene chromium tricarbonyl

24-0120-1G 24-0120-5G
100211-038EA 196.74 USD
100211-038 100211-040
Chromium benzene tricarbonyl ≥98%
Chromium benzene tricarbonyl
Formula: (C₆H₆)Cr(CO)₃
MW: 214.14 g/mol
Melting Pt: 165…166 °C
MDL Number: MFCD00009955
CAS Number: 12082-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR